Anti-SURF1 Rabbit Polyclonal Antibody
Catalog # PRSI22-466
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:SURF1 cytochrome c oxidase assembly factor
- Antigen symbol:SURF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6834
- Antigen synonyms:surfeit locus protein 1|surfeit 1|CMT4K
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 80-275 of human SURF1 (NP_003163.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SURF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat