Anti-RPA3 Rabbit Polyclonal Antibody
Catalog # PRSI22-436
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Replication protein A3, 14kDa
- Antigen symbol:RPA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6119
- Antigen synonyms:Replication protein A3 (14kD)|RF-A protein 3|RPA3|RP-A p14|RPA14|Replication factor A protein 3|14kDa|REPA3|Replication protein A3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 6-121 of human RPA3 (NP_002938.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RPA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat