Anti-HGD Rabbit Polyclonal Antibody
PRSI22-356
New Product
- Antibody type:Primary
- Antigen name:Homogentisate 1,2-dioxygenase
- Antigen symbol:HGD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3081
- Antigen synonyms:homogentisic acid oxidase|AKU|homogentisicase|HGO|homogentisate oxygenase|homogentisate 1,2-dioxygenase|homogentisate oxidase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 226-445 of human HGD (NP_000178.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: HGD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat