Anti-CST2 Rabbit Polyclonal Antibody
PRSI22-322
New Product
- Antibody type:Primary
- Antigen name:Cystatin SA
- Antigen symbol:CST2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1470
- Antigen synonyms:cystatin-SA|cysteine-proteinase inhibitor|salivary cysteine (thiol) protease inhibitor|cystatin S5|cystatin 2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-141 of human CST2 (NP_001313.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse