Anti-HSD3B2 Rabbit Polyclonal Antibody
PRSI18-323
- Antibody type:Primary
- Antigen name:Hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2
- Antigen symbol:HSD3B2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3284
- Antigen synonyms:HSDB|SDR11E2|HSD3B|HSDB3|HSD3B2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human HSD3B2 (NP_000189,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: HSD3B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse