Anti-LBP Rabbit Polyclonal Antibody
PRSI16-962
New Product
- Antibody type:Primary
- Antigen name:Lipopolysaccharide binding protein
- Antigen symbol:LBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:3929
- Antigen synonyms:BPIFD2|MGC22233|LBP
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein of Human LBP
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: LBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat