Anti-MCU Rabbit Polyclonal Antibody
PRSI16-524
New Product
- Antibody type:Primary
- Antigen name:Mitochondrial calcium uniporter
- Antigen symbol:MCU
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:90550
- Antigen synonyms:CCDC109A|mitochondrial|C10orf42|coiled-coil domain-containing protein 109A|calcium uniporter protein|HsMCU
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human MCU (NP_612366,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MCU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat