Anti-SPARC Rabbit Polyclonal Antibody
Catalog # PRSI16-455
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:SPARC
- Antigen symbol:SPARC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6678
- Antigen synonyms:Basement-membrane protein 40|ON|SPARC|Osteonectin|BM-40|Secreted protein acidic and rich in cysteine
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 18-303 of human SPARC (NP_003109,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat