Anti-AZIN2 Rabbit Polyclonal Antibody
Catalog # PRSI16-332
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antizyme inhibitor 2
- Antigen symbol:AZIN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:113451
- Antigen synonyms:ODCp|AZI2|AZIB1|ornithine decarboxylase like|ADC|ODC-p|ornithine decarboxylase-like protein|ODC1L|arginine decarboxylase|ODC antizyme inhibitor-2|ODC-paralogue|ODC-like protein|antizyme inhibitor 2|ornithine decarboxylase-paralog
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-360 of human AZIN2 (NP_443724,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: AZIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse