Anti-TCF19 Rabbit Polyclonal Antibody
Catalog # PRSI22-032
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor 19
- Antigen symbol:TCF19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6941
- Antigen synonyms:TCF19|SC1|transcription factor 19 (SC1)
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human TCF19 (NP_009040.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TCF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat