Anti-ORF3a Sheep Polyclonal Antibody
Catalog # PRSI20-282
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:ORF3a protein
- Antigen symbol:ORF3a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Reactivity:Virus
- Western blot:Yes
- Form:Liquid
- Gene ID:4374569
- Antigen synonyms:SARS-CoV-2-ORF3A
- Storage buffer:Antibody supplied in PBS.
- Molecular weight:58 kDa
- Storage temperature:Aliquot and store at −20...+730 °C. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:Batch dependent
- Shipping temperature:Dry Ice
- Immunogen:GST-SARS-CoV2 ORF3A .
- Purification:Antibodies are affinity purified against the antigen and eluted with low pH.
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ORF3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Virus