Anti-MAGOH Rabbit Polyclonal Antibody
PRSI19-984
New Product
- Antibody type:Primary
- Antigen name:Mago-nashi homolog, proliferation-associated (Drosophila)
- Antigen symbol:MAGOH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4116
- Antigen synonyms:proliferation-associated (Drosophila)|MAGOH|mago-nashi homolog|MAGOHA
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-146 of human MAGOH (NP_002361.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MAGOH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat