Anti-NUDT1 Rabbit Polyclonal Antibody
PRSI19-686
New Product
- Antibody type:Primary
- Antigen name:Nudix hydrolase 1
- Antigen symbol:NUDT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4521
- Antigen synonyms:8-oxo-dGTPase|8-oxo-7,8-dihydroguanosine triphosphatase|2-hydroxy-dATP diphosphatase|8-oxo-7,8-dihydrodeoxyguanosine triphosphatase|nucleoside diphosphate-linked moiety X motif 1|nucleoside diphosphate-linked moiety X-type motif 1|7,8-dihydro-8-oxoguanine triphosphatase|mutT human homolog 1|nudix motif 1|MTH1|nudix (nucleoside diphosphate linked moiety X)-type motif 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human NUDT1 (NP_945192.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NUDT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat