Anti-SIGLEC9 Rabbit Polyclonal Antibody
PRSI19-433
New Product
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 9
- Antigen symbol:SIGLEC9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:27180
- Antigen synonyms:SIGLEC9|CDw329|FOAP-9|siglec-9|OBBP-LIKE|CD329
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 210-340 of human SIGLEC9 (NP_001185487.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SIGLEC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat