Anti-SLC1A4 Rabbit Polyclonal Antibody
Catalog # PRSI18-934
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 1 (glutamate/neutral amino acid transporter), member 4
- Antigen symbol:SLC1A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6509
- Antigen synonyms:SATT|ASCT1|SLC1A4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human SLC1A4.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC1A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat