Anti-EFNA3 Rabbit Polyclonal Antibody
Catalog # PRSI18-788
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ephrin A3
- Antigen symbol:EFNA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1944
- Antigen synonyms:Ehk1-L|eph-related receptor tyrosine kinase ligand 3|LERK-3|EHK1 ligand|EFL2|EPLG3|LERK3|ephrin-A3|EFL-2|ligand of eph-related kinase 3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 23-214 of human EFNA3 (NP_004943.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: EFNA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse