Anti-CAPN2 Rabbit Polyclonal Antibody
PRSI18-761
New Product
- Antibody type:Primary
- Antigen name:Calpain 2
- Antigen symbol:CAPN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:824
- Antigen synonyms:CANP2|(m/II) large subunit|CANP 2|calpain|large subunit|calpain-2 catalytic subunit|CANPL2|calpain M-type|mCANP|large polypeptide L2|large [catalytic] subunit|calcium-activated neutral proteinase 2|millimolar-calpain|M-calpain|calpain 2|CANPml
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of human CAPN2.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CAPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat