Anti-DCAF12L2 Rabbit Polyclonal Antibody
PRSI60-121
New Product
- Antibody type:Primary
- Antigen name:DDB1- and CUL4-associated factor 12-like protein 2
- Antigen symbol:DCAF12L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:312 - 338 aa
- Form:Liquid
- Gene ID:340578
- Modification:None
- Molecular weight:51 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,4 mg/ml
- Shipping temperature:4°C
- Immunogen:This DCAF12L2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 312 - 338 amino acids from the C-terminal region of human DCAF12L2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: DCAF12L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 312-338
Host: Rabbit
Isotype: Ig
Reactivity: Human