Order Entry
United Kingdom
ContactUsLinkComponent
Anti-PPP1R1C Rabbit Polyclonal Antibody
Anti-PPP1R1C Rabbit Polyclonal Antibody
Catalog # PRSI60-075
Supplier:  ProSci Inc.
New Product
undefined
Anti-PPP1R1C Rabbit Polyclonal Antibody
Catalog # PRSI60-075
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein phosphatase 1 regulatory subunit 1C
  • Antigen symbol:
    PPP1R1C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    50 - 76 aa
  • Form:
    Liquid
  • Gene ID:
    151242
  • Modification:
    None
  • Molecular weight:
    12 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4°C
  • Immunogen:
    This PPP1R1C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 50 - 76 amino acids from the N-terminal region of human PPP1R1C.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: PPP1R1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 50-76
Host: Rabbit
Isotype: Ig
Reactivity: Mouse