Anti-KIT Rabbit Polyclonal Antibody
PRSI60-070
New Product
- Antibody type:Primary
- Antigen name:Mast/stem cell growth factor receptor Kit
- Antigen symbol:KIT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:708 - 737 aa
- Form:Liquid
- Gene ID:3815
- Modification:None
- Molecular weight:110 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This KIT antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 708 - 737 amino acids from the Central region of human KIT.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: KIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 708-737
Host: Rabbit
Isotype: Ig
Reactivity: Human