Anti-OR2AG2 Rabbit Polyclonal Antibody
PRSI59-790
New Product
- Antibody type:Primary
- Antigen name:Olfactory receptor 2AG2
- Antigen symbol:OR2AG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:64 - 90 aa
- Form:Liquid
- Gene ID:338755
- Modification:None
- Molecular weight:35 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This OR2AG2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 64 - 90 amino acids from the N-terminal region of human OR2AG2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: OR2AG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 64-90
Host: Rabbit
Isotype: Ig
Reactivity: Human