Anti-RARB Rabbit Polyclonal Antibody
PRSI59-680
New Product
- Antibody type:Primary
- Antigen name:Retinoic acid receptor beta
- Antigen symbol:RARB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:168 - 195 aa
- Form:Liquid
- Gene ID:5915
- Modification:None
- Molecular weight:50 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This RARB antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 168 - 195 amino acids from the Central region of human RARB.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: RARB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 168-195
Host: Rabbit
Isotype: Ig
Reactivity: Human