Anti-VDAC3 Rabbit Polyclonal Antibody
Catalog # PRSI58-941
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Voltage-dependent anion-selective channel protein 3
- Antigen symbol:VDAC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:156 - 183 aa
- Form:Liquid
- Gene ID:7419
- Modification:None
- Molecular weight:31 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,29 mg/ml
- Shipping temperature:4°C
- Immunogen:This VDAC3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 156 - 183 amino acids from the Central region of human VDAC3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: VDAC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 156-183
Host: Rabbit
Isotype: Ig
Reactivity: Human