Anti-SLC27A3 Rabbit Polyclonal Antibody
Catalog # PRSI57-055
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Long-chain fatty acid transport protein 3
- Antigen symbol:SLC27A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:124-152 aa
- Form:Liquid
- Gene ID:11000
- Modification:None
- Molecular weight:79 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SLC27A3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 124-152 amino acids from the N-terminal region of human SLC27A3
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: SLC27A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 124-152
Host: Rabbit
Isotype: Ig
Reactivity: Human