Anti-Csf1r Rat Monoclonal Antibody [clone: AFS98] (FITC (Fluorescein Isothiocyanate))
PRSI76-878
New Product
- Antibody type:Primary
- Antigen name:colony stimulating factor 1 receptor
- Antigen symbol:Csf1r
- Clonality:Monoclonal
- Clone:AFS98
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P09581
- Antigen synonyms:Csf1r
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
The AFS98 monoclonal antibody specifically reacts with the mouse CD115 molecule, a 150 kDa receptor for the colony stimulating factor (CSF-1) or macrophage CFS (M-CFS), expressed by some epithelial cells, monocytes, osteoclasts, and macrophages. The colony stimulating factor-1 regulates the proliferation and the differentiation of the monocytic lineage cells. The AFS98 antibody can be used to identify myeloid lineage cells.
Type: Primary
Antigen: Csf1r
Clonality: Monoclonal
Clone: AFS98
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse