Anti-CDH5 Mouse Monoclonal Antibody [clone: 16B1] (PE (Phycoerythrin))
PRSI76-861
New Product
- Antibody type:Primary
- Antigen name:cadherin 5, type 2 (vascular endothelium)
- Antigen symbol:CDH5
- Clonality:Monoclonal
- Clone:16B1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Cross adsorption:No
- Form:liquid
- Gene ID:P33151
- Antigen synonyms:CDH5
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
The 16B1 monoclonal antibody specifically reacts with human CD144, the 140 kDa molecule called vascular endothelial (VE-cadherin) or cadherin 5. CD144 is an endothelial specific calcium-dependent adhesion molecule involved in cell contact-dependent growth inhibition, migration, survival, adhesion. It is concentrated at the intercellular boundaries of endothelial cells and essential in maintaining cell layer integrity.
Type: Primary
Antigen: CDH5
Clonality: Monoclonal
Clone: 16B1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human