Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
PRSI76-766
New Product
- Antibody type:Primary
- Antigen name:CD19 molecule
- Antigen symbol:CD19
- Clonality:Monoclonal
- Clone:HIB19
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P15391
- Antigen synonyms:CD19
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:25 Tests
The HIB19 monoclonal antibody reacts with a human 95 kDa transmembrane glycoprotein known as CD19, which is expressed by B lymphocytes during all the developmental stages, except for the terminally differentiated plasma cells. CD19 is also expressed on follicular dendritic cells, and seems to ensure the regulation of B lymphocytes proliferation. CD19, CD21, CD81, MHC class II, and Leu13 can bind together and form a complex which associates with the B cell receptor (BCR) on the surface of B lymphocytes and facilitates the signal transduction.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human