Anti-KLRB1 Mouse Monoclonal Antibody [clone: HP-3G10] (FITC (Fluorescein Isothiocyanate))
PRSI76-835
New Product
- Antibody type:Primary
- Antigen name:killer cell lectin-like receptor subfamily B, member 1
- Antigen symbol:KLRB1
- Clonality:Monoclonal
- Clone:HP-3G10
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:Q12918
- Antigen synonyms:KLRB1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:25 Tests
The HP-3G10 monoclonal antibody specifically binds to an 80 kDa homodimer type II membrane glycoprotein from the C-type lectin superfamily, known as the human CD161 or NKR-P1A. CD161 is expressed on most natural killer cells, subsets of CD4+ and CD8+ T lymphocytes, gamma delta TCR T lymphocytes, a subset of CD3+ thymocytes, and especially on CD45RO+ T lymphocytes. Reports indicate that it may serve as a specific receptor for some natural killer cell targets and a possible stimulatory role.
Type: Primary
Antigen: KLRB1
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human