Anti-FCGR3A Mouse Monoclonal Antibody [clone: CB16] (FITC (Fluorescein Isothiocyanate))
PRSI76-570
New Product
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
- Antigen symbol:FCGR3A
- Clonality:Monoclonal
- Clone:CB16
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P08637
- Antigen synonyms:FCGR3A
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
The CB16 monoclonal antibody specifically reacts with human CD16, the low affinity IgG receptor III (FC gamma RIII). CD16 is expressed on granulocytes, monocytes, macrophages, and NK cells and plays a role in NK activation and signal transduction. CD16 is expressed as either CD16a or CD16b. CD16a is a polypeptide-anchored transmembrane protein and CD16b is a glycosylphosphatidylinositol (GPI)-anchored protein that is expressed exclusively on neutrophils.
Type: Primary
Antigen: FCGR3A
Clonality: Monoclonal
Clone: CB16
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human