Specifications
- Antibody type:Primary
- Antigen name:CD19 antigen
- Antigen symbol:Cd19
- Clonality:Monoclonal
- Clone:1D3
- Conjugation:VioletFluor® 450
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P25918
- Antigen synonyms:Cd19
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
Specifications
About this item
The 1D3 monoclonal antibody specifically reacts with mouse CD19, a 95 kDa transmembrane glycoprotein, a member of the Ig superfamily and a B cell-lineage differentiation antigen expressed by all the B lymphocyte development stages, except for the terminally differentiated plasma cells. CD19 associates with CD21, CD81 and MHC class II to form a multi-molecular complex that initiates the mature B lymphocyte activation by interaction with the B-cell receptors. CD 19 enhances the B cell proliferation, development and activation, and the maturation of memory B cells. In CD19-deficient mice, the generation and maturation of B lymphocytes in the bone marrow and periphery are affected. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450 nm.
Type: Primary
Antigen: Cd19
Clonality: Monoclonal
Clone: 1D3
Conjugation: VioletFluor® 450
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse