Anti-SIGLEC7 Rabbit Polyclonal Antibody
Catalog # PRSI58-414
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid-binding Ig-like lectin 7
- Antigen symbol:SIGLEC7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:407 - 438 aa
- Form:Liquid
- Gene ID:27036
- Modification:None
- Molecular weight:51 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SIGLEC7 (D-siglec) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 407-438 amino acids from the C-terminal region of human SIGLEC7 (D-siglec)
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: SIGLEC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 407-438
Host: Rabbit
Isotype: Ig
Reactivity: Human