Anti-SIGLEC9 Rabbit Polyclonal Antibody
PRSI58-379
New Product
- Antibody type:Primary
- Antigen name:Sialic acid-binding Ig-like lectin 9
- Antigen symbol:SIGLEC9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:385 - 415 aa
- Form:Liquid
- Gene ID:27180
- Modification:None
- Molecular weight:50 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SIGLEC9 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 385-415 amino acids from the C-terminal region of human SIGLEC9
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: SIGLEC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 385-415
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse