Anti-SIGLEC8 Rabbit Polyclonal Antibody
Catalog # PRSI58-370
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid-binding Ig-like lectin 8
- Antigen symbol:SIGLEC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:373 - 406 aa
- Form:Liquid
- Gene ID:27181
- Modification:None
- Molecular weight:54 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1,67 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SIGLEC8 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 373-406 amino acids from the C-terminal region of human SIGLEC8
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: SIGLEC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 373-406
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse