Anti-DKK3 Rabbit Polyclonal Antibody
Catalog # PRSI58-169
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Dickkopf-related protein 3
- Antigen symbol:DKK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:15 - 45 aa
- Form:Liquid
- Gene ID:27122
- Modification:None
- Molecular weight:38 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DKK3 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 15-45 amino acids from the N-terminal region of human DKK3
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: DKK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 15-45
Host: Rabbit
Isotype: Ig
Reactivity: Human