Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Tlr2 Rabbit Polyclonal Antibody
Anti-Tlr2 Rabbit Polyclonal Antibody
Catalog # PRSI58-160
Supplier:  ProSci Inc.
New Product
undefined
Anti-Tlr2 Rabbit Polyclonal Antibody
Catalog # PRSI58-160
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like receptor 2
  • Antigen symbol:
    Tlr2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    720 - 749 aa
  • Form:
    Liquid
  • Gene ID:
    24088
  • Modification:
    None
  • Molecular weight:
    89 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Mouse TLR2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 720-749 amino acids from the C-terminal region of mouse TLR2
  • Purification:
    This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: Tlr2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 720-749
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse