Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CASP12 Rabbit Polyclonal Antibody
Anti-CASP12 Rabbit Polyclonal Antibody
Catalog # PRSI13-082
Supplier:  ProSci Inc.
New Product
Anti-CASP12 Rabbit Polyclonal Antibody
Catalog # PRSI13-082
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    caspase 12 (gene/pseudogene)
  • Antigen symbol:
    CASP12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    100506742
  • Antigen synonyms:
    OTTHUMP00000207028|caspase 12 pseudogene 1|OTTHUMP00000219058|OTTHUMP00000207029|OTTHUMP00000207031|inactive caspase-12|CASP12P1|caspase 12 (gene/pseudogene)|OTTHUMP00000207026|OTTHUMP00000207032|CASP12|CASP-12
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CASP12 (NP_001177945.2)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CASP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat