Anti-ASIC2 Rabbit Polyclonal Antibody
Catalog # PRSI63-841
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Acid-sensing ion channel 2
- Antigen symbol:ASIC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:120-148 aa
- Form:Liquid
- Gene ID:40
- Modification:None
- Molecular weight:58 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,28 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ACCN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 120-148 amino acids from the Central region of human ACCN1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ASIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 120-148
Host: Rabbit
Isotype: Ig
Reactivity: Human