Anti-CYP4F3 Rabbit Polyclonal Antibody
Catalog # PRSI63-566
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Docosahexaenoic acid omega-hydroxylase CYP4F3
- Antigen symbol:CYP4F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:77-109 aa
- Form:Liquid
- Gene ID:4051
- Modification:None
- Molecular weight:60 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CYP4F3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 77-109 amino acids from the N-terminal region of human CYP4F3.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: CYP4F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 77-109
Host: Rabbit
Isotype: Ig
Reactivity: Human