Anti-EIF4B Rabbit Polyclonal Antibody
PRSI60-584
New Product
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 4B
- Antigen symbol:EIF4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:353 - 382 aa
- Form:Liquid
- Gene ID:1975
- Modification:None
- Molecular weight:69 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This EIF4B antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 353 - 382 amino acids from the Central region of human EIF4B.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: EIF4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 353-382
Host: Rabbit
Isotype: Ig
Reactivity: Human