Anti-ST6GAL2 Rabbit Polyclonal Antibody
Catalog # PRSI60-558
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Beta-galactoside alpha-2,6-sialyltransferase 2
- Antigen symbol:ST6GAL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:483 - 509 aa
- Form:Liquid
- Gene ID:84620
- Modification:None
- Molecular weight:60 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,37 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ST6GAL2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 483 - 509 amino acids from the C-terminal region of human ST6GAL2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ST6GAL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 483-509
Host: Rabbit
Isotype: Ig
Reactivity: Human