Anti-DIMT1 Rabbit Polyclonal Antibody
PRSI60-554
New Product
- Antibody type:Primary
- Antigen name:Probable dimethyladenosine transferase
- Antigen symbol:DIMT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:268 - 298 aa
- Form:Liquid
- Gene ID:27292
- Modification:None
- Molecular weight:35 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DIMT1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 268 - 298 amino acids from the C-terminal region of human DIMT1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: DIMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 268-298
Host: Rabbit
Isotype: Ig
Reactivity: Human