Anti-HAS2 Rabbit Polyclonal Antibody
Catalog # PRSI23-964
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:hyaluronan synthase 2
- Antigen symbol:HAS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:3037
- Antigen synonyms:hyaluronate synthase 2|HA synthase 2|hyaluronan synthase 2|hyaluronic acid synthase 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 67-185 of human HAS2 (NP_005319.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: HAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse