Anti-ADAM33 Rabbit Polyclonal Antibody
PRSI23-483
New Product
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 33
- Antigen symbol:ADAM33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:80332
- Antigen synonyms:MGC71889|DJ964F7.1|FLJ35308|MGC149823|C20orf153|DKFZp434K0521|ADAM33|FLJ36751
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human ADAM33 (NP_001269376.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ADAM33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat