Anti-Timd4 Recombinant Antibody [Clone: 3A1]
Catalog # PRSI24-620
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Timd4
- Antigen symbol:Timd4
- Clonality:Recombinant
- Clone:3A1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:276891
- Antigen synonyms:Spleen|TIMD4|T-cell immunoglobulin and mucin domain containing 4|knockout of lymphotoxin protein|TIMD-4|T-cell membrane protein 4|T-cell immunoglobulin and mucin domain-containing protein 4|SMUCKLER|TIM4|mucin-containing|TIM-4|T-cell immunoglobulin mucin receptor 4
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising female Lewis rats (Harlan Sprague-Dawley) with Tim-4:Ig fusion protein in complete Freund's adjuvant
- Tested applications:Blocking, therapeutics
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: Timd4
Clonality: Recombinant
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Mouse