Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SLC3A2 Recombinant Antibody [Clone: CP1-5]
Anti-SLC3A2 Recombinant Antibody [Clone: CP1-5]
Catalog # PRSI24-348
Supplier:  ProSci Inc.
Anti-SLC3A2 Recombinant Antibody [Clone: CP1-5]
Catalog # PRSI24-348
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 3 member 2
  • Antigen symbol:
    SLC3A2
  • Clonality:
    Recombinant
  • Clone:
    CP1-5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    6520
  • Antigen synonyms:
    large neutral amino acid transporter|LAT1|oncoprotein CD98
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Epitope peptide (LLHGDFHAFSAG-GGGCCD) linked to KLH
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: solute carrier family 3 member 2
Clonality: Recombinant
Clone: CP1-5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human