Anti-EEC1 Recombinant Antibody [Clone: BBM.1]
Catalog # PRSI24-234
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:ectrodactyly, ectodermal dysplasia and cleft lip/palate syndrome 1
- Antigen symbol:EEC1
- Clonality:Recombinant
- Clone:BBM.1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:1913
- Antigen synonyms:CDABP0092|HDCMA22P|Beta-2-microglobulin|BBM1|B2M
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Molt 4, a human T cell line
- Tested applications:Depletion
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: ectrodactyly, ectodermal dysplasia and cleft lip/palate syndrome 1
Clonality: Recombinant
Clone: BBM.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Human