Anti-GLB1 Recombinant Antibody [Clone: 13R4]
PRSI24-152
- Antibody type:Primary
- Antigen name:galactosidase beta 1
- Antigen symbol:GLB1
- Clonality:Recombinant
- Clone:13R4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG1 lambda
- Reactivity:E. coli
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:2720
- Antigen synonyms:MPS4B| Acid beta galactosidase|EBP|β-galactosidase|β-gal|GLB1|beta-gal|ELNR1|Galactosidase beta 1|Beta galactosidase 1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Beta-galactosidase
- Tested applications:Negative control
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: galactosidase beta 1
Clonality: Recombinant
Clone: 13R4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 lambda
Reactivity: E. coli