Anti-CD33 Recombinant Antibody [Clone: M195]
Catalog # PRSI31-535
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD33 molecule
- Antigen symbol:CD33
- Clonality:Recombinant
- Clone:M195
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:945
- Antigen synonyms:gp67|Siglec-3|Myeloid cell surface antigen CD33|Sialic acid-binding Ig-like lectin 3
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The original antibody was generated by immunizing mice with live leukemic myeloblasts
- Tested applications:RIA,Therapeutics
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD33 molecule
Clonality: Recombinant
Clone: M195
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human