Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

1
Catalog # SERA37296.01
Marketsource
Supplier:   Serva
Trypsin
Catalog # SERA37296.01
Supplier:  Serva
Marketsource
CAS Number:  
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Cat. No.:
    SERA37296.01
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00082094
  • Grade:
    GMP
  • Enzyme name:
    Trypsin
  • CAS No.:
    9002-07-7

Specifications