Phenol red sodium salt
Catalog # SERA32097.02
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:376.35 g/mol
Merck Index:12,07397
Melting Pt:285 °C
Storage Temperature:Ambient
EINECS:252-057-8
CAS Number:34487-61-1
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901